Imprimer
GF Signet Metalex Paddle-Wheel Flow Sensors
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
05618-60 05618-64
MFLX05618-60EA 965 EUR
MFLX05618-60 MFLX05618-64
GF Signet Metalex Paddle-Wheel Flow Sensors
Débitmètres
Stainless steel body withstands high pressures and temperatures.
  • Use in agressive liquid flow applications
Use these rugged sensors to monitor the flow of liquid ammonia, turbine steam condensate, and more—wherever strength and chemical resistance are required.
Open-core rotor is cavitation free at flow rates from 1.5 to 20 ft/second. Easy maintenance—sensors can be replaced in minutes.
METALEX flow sensors are compatible with all of our battery- and line-powered flowmeters, accumulators, and controllers. Sensors must be used with the fitting type specified in the table below—installation fittings sold separately.
METALEX sensors feature CD4MCU rotor; Fluoroloy B rotor bearings; and 316 stainless steel body, rotor shaft, and shaft retainers.
Informations de livraison: Includes 25-foot signal cable.