Imprimer
Masterflex® Direct-Read Variable Area Flowmeters for Water, Dual-Scale, Stainless Steel, Avantor®
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
32205-08 32205-12 32205-14
MFLX32205-02EA 1350 EUR
MFLX32205-02 MFLX32205-08 MFLX32205-12 MFLX32205-14
Masterflex® Direct-Read Variable Area Flowmeters for Water, Dual-Scale, Stainless Steel, Avantor®
Débitmètres
Install in any position.
  • Direct-reading flowmeters feature a calibrated scale for a specific medium - no need to convert readings to get flow rate│Use with water for laboratory and industrial applications
Dual-scale flowmeters indicate gallons and liters simultaneously with an easy-to-read linear scale and highly visible red indicator.