Imprimer
Masterflex® Direct-Reading Variable Area Flowmeters for Water, Brass Housing, Avantor®
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
32204-06 32204-14 32204-16 32204-22 32204-24
MFLX32204-04EA 362 EUR
MFLX32204-04 MFLX32204-10 MFLX32204-06 MFLX32204-14 MFLX32204-16 MFLX32204-22 MFLX32204-24
Masterflex® Direct-Reading Variable Area Flowmeters for Water, Brass Housing, Avantor®
Débitmètres
Don’t sacrifice suitability for precision.
  • Direct-reading flowmeters feature a calibrated scale for a specific medium - no need to convert readings to get flow rate│Use with water for laboratory and industrial applications
Use for high pressure industrial applications—install in any configuration, even next to a bend in your pipeline. A calibrated retention spring eliminates the need to mount flowmeters vertically or run straight lengths of pipe prior to or after the meter. These rugged flowmeters are relatively insensitive to shock and vibrations—the spring, piston assembly, and internal magnet compensate for rough process environments.