Imprimer
CPC® High-Flow Quick-Disconnect Fittings, Threaded Inserts
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
31308-72BLK
MFLX31303-30EA 10.9 EUR
MFLX31303-30 MFLX31303-31 MFLX31303-35 MFLX31303-36 MFLX31303-37 MFLX31307-35 MFLX31307-36 MFLX31307-37 MFLX31303-37BLK MFLX31307-37BLK MFLX31303-35BLK MFLX31303-36BLK MFLX31307-35BLK MFLX31303-31BLK MFLX31303-30BLK MFLX31307-36BLK MFLX31303-32BLK MFLX31308-72BLK
CPC® High-Flow Quick-Disconnect Fittings, Threaded Inserts
Raccords et connecteurs
No more leaks - valved couplings automatically shut off flow.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Compact, lightweight couplings feature a high flow and low spillage design
These CPC® high-flow quick-disconnect fittings have flow comparable to many ¹/₂" flow couplings in a ³/₈" body size. An ergonomic design and a large, shrouded thumb latch pad produce a coupling that is easy to grip and simple to operate. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. To disconnect fitting halves, simply press the button on the coupling body. Valved models have a spring-operated 316 stainless steel valve that seals the flow path when fitting halves are disengaged. All fitting bodies have 316 stainless steel latch springs and EPDM seals.
For a complete coupling, order one fitting body and one insert.