Imprimer
Union Fittings, Male Threaded Adapter Nipple, Straight
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
40610-90 40610-91 40610-92 40610-93 40610-94 40640-90 06546-22 06546-24 06546-26 06546-29 40610-95 40615-23 40615-25 40615-26 40615-27 40615-28
MFLX30623-20EA 20.7 EUR
MFLX30623-20 MFLX30623-21 MFLX30623-24 MFLX30623-25 MFLX30623-26 MFLX30623-27 MFLX30623-28 MFLX30623-29 MFLX30623-31 MFLX30627-28 MFLX30629-02 MFLX40610-90 MFLX40610-91 MFLX40610-92 MFLX40610-93 MFLX40610-94 MFLX40640-90 MFLX06546-22 MFLX06546-24 MFLX06546-26 MFLX06546-29 MFLX40610-95 MFLX30627-24 MFLX30627-25 MFLX40615-23 MFLX40615-25 MFLX40615-26 MFLX40615-27 MFLX40615-28
Union Fittings, Male Threaded Adapter Nipple, Straight
Raccords et connecteurs
Maintain a permanent connection.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Threaded fittings provide a more permanent fluid path. Choose the size and material to meet your application needs.