Imprimer
Masterflex® Adapter Fittings, Hose Barb to Male UNF Threaded, Avantor®
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
40614-04
MFLX31209-40EA 22.3 EUR
MFLX31209-40 MFLX31209-41 MFLX31501-53 MFLX31501-54 MFLX31501-55 MFLX40621-00 MFLX40621-02 MFLX40621-03 MFLX40621-05 MFLX40621-06 MFLX40625-00 MFLX40625-05 MFLX40629-00 MFLX41517-04 MFLX41517-05 MFLX50621-01 MFLX50621-04 MFLX50621-07 MFLX50625-04 MFLX51517-03 MFLX51517-11 MFLX30704-84 MFLX40625-02 MFLX40625-06 MFLX41517-09 MFLX50625-01 MFLX40614-04 MFLX40614-00 MFLX40614-02 MFLX40614-01 MFLX40614-91 MFLX40625-03 MFLX40629-01
Masterflex® Adapter Fittings, Hose Barb to Male UNF Threaded, Avantor®
Raccords et connecteurs
No tools needed for assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple and economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).