Imprimer
Stainless steel lockers
Fournisseur: TEKNOMEK
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
CRLK-DLSSF CRLK-DLSSS CRLK-MDSS110 CRLK-MDSS220 LKD300-1 LKD300-2 LKD300-3 LKD300-4 LKS300-1 LKS300-2 LKS300-3 LKS300-4 LKS450-1 LKS450-2 LKS450-3 LKS450-4
TEKMCRLK-DLSSFEA 0 EUR
TEKMCRLK-DLSSF TEKMCRLK-DLSSS TEKMCRLK-MDSS110 TEKMCRLK-MDSS220 TEKMLKD300-1 TEKMLKD300-2 TEKMLKD300-3 TEKMLKD300-4 TEKMLKS300-1 TEKMLKS300-2 TEKMLKS300-3 TEKMLKS300-4 TEKMLKS450-1 TEKMLKS450-2 TEKMLKS450-3 TEKMLKS450-4
Stainless steel lockers
Mobilier Casiers de vestiaires
These intelligently designed lockers are supplied fully welded and ready to use.
  • High-quality 304 grade stainless steel
  • Fully welded, ready to use
  • Camlock as standard
  • Easy clean doors
  • Tough and robust design
  • Sloping top for improved hygiene
Using 304 grade stainless steel, they feature integral legs, 150 mm from the floor to allow for easy clean access with stainless steel levelling feet for stability. Choose from dirty laundry lockers, to multi-door lockers for small items to lockers with more space for personal possessions.