Imprimer
Storage cupboards with inset sink
Fournisseur: Teknomek
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
134-0511EA 0 EUR
134-0511 134-0512 134-0510
Storage cupboards with inset sink
Robinets et éviers Éviers
Stainless steel sink with double cupboard and drawer is manufactured from high quality, 304 grade stainless steel and is supplied with an inset sink and an in-built storage cupboard for ultimate versatility.
  • Fully welded 304 grade stainless steel
  • Brushed dull polish finish
  • Inset stainless steel basin
  • Stainless steel adjustable feet with non-marking rubber base
  • Stainless steel adjustable shelf
  • Magnetic catches on doors
  • Lever taps included
  • Rear upstand
The 1200 and 1500 mm wide cupboards have a drawer too!