Imprimer
VWR®, Septa 9,5 mm
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
548-3327AEA 170 EUR
548-3327A 548-3292A 548-3328A 548-3329A 548-3186A 548-3258A
VWR®, Septa 9,5 mm
Septa
Almost all septa have one laminated side with PTFE (high chemical resistance) and form an inert barrier between carrier material and sample.
  • Fits for a broad range of caps
  • Easy exchange
  • Different hardness and septa thickness available
Septa choice depends on the application.