Imprimer
VWR® Septa ND8
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
548-0027AEA 53.4 EUR
548-0027A 548-0312A 548-0320A 548-0316A 548-0318A 548-3344A 548-1460A 548-0313A 548-0314A 548-0026A 548-0828A 548-0315A
VWR® Septa ND8
Septa
Almost all septa have one laminated side with PTFE (high chemical resistance) and form an inert barrier between carrier material and sample.
  • Fits for a broad range of caps
  • Easy exchange
  • Different hardness and septa thickness available
Septa choice depends on the application.