Imprimer
VWR®, Screw Neck Vials ND8
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
548-0019AEA 349 EUR
548-0019A 548-0175A 548-2124A 548-0914A 548-1862A 548-0176A 548-0095A 548-0850A 548-2090A 548-0018A 548-0420A 548-0448A 548-0819A 548-0853A 548-0913A 548-0915A 548-1474A 548-1517A 548-1420A 548-1878A 548-0419A 548-1863A 548-2587A 548-2588A
VWR®, Screw Neck Vials ND8
Fioles
ND8 screw neck vials are manufactured from 1st hydrolytic class borosilicate glass with flat bottom. They can be purchased with a variety of polypropylene seal styles and septa materials.
  • High performance caps/septa
  • Good handling with gloves
  • Quick assembly
  • Small opening
ND8 screw neck vials are constructed from type I borosilicate glass with low metal content. Vials are available in amber or clear versions with a flat bottom to ensure compatibility with inserts. Associated screw caps, mostly available with assembled septa, are engineered to prevent sample contamination while providing consistent, secure seals. The use of closed caps allows the samples to be stored for a long time (long-term storage applications).