Imprimer
VWR®, Septa 18 mm
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
548-3223AEA 681 EUR
548-3223A 548-0806A 548-0866A 548-1464A 548-0867A 548-3224A 548-3221A 548-3315A
VWR®, Septa 18 mm
Septa
Almost all septa have one laminated side with PTFE (high chemical resistance) and form an inert barrier between carrier material and sample.
  • Fits for a broad range of caps
  • Easy exchange
  • Different hardness and septa thickness available
Septa choice depends on the application.