Imprimer
VWR®, Septa 12 mm
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
548-0480AEA 33.6 EUR
548-0480A 548-0111A 548-0476A 548-0108A 548-0475A 548-0112A 548-0479A 548-0477A
VWR®, Septa 12 mm
Septa
Almost all septa have one laminated side with PTFE (high chemical resistance) and form an inert barrier between carrier material and sample.
  • Fits for a broad range of caps
  • Easy exchange
  • Different hardness and septa thickness available
Septa choice depends on the application.