Imprimer
ESD Safety Shoes, DANIEL Pro, low, S3
Fournisseur: ELTEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1727901/36 1727901/37 1727901/38 1727901/39 1727902/36 1727902/37 1727902/38 1727902/39 1727903/36 1727903/37 1727903/38 1727903/39
ELTN1727901/36EA 91.9 EUR
ELTN1727901/36 ELTN1727901/37 ELTN1727901/38 ELTN1727901/39 ELTN1727902/36 ELTN1727902/37 ELTN1727902/38 ELTN1727902/39 ELTN1727903/36 ELTN1727903/37 ELTN1727903/38 ELTN1727903/39
ESD Safety Shoes, DANIEL Pro, low, S3
Chaussures et bottes
This shoe is ideally tailored to the ergonomics of female feet. Excellent wearing comfort, with a padded upper edge to protect the Achilles tendon.
  • Hydrophobised nubuck leather
  • Fabric lining for natural ventilation
  • Full, padded bellows tongue
  • Semi-orthopaedic inlay sole ESD black
  • Non-metallic, penetration-resistant midsole
  • PU/PU sole ERGO-ACTIVE 2
  • TPU scuff cap and steel toe cap
Excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA).