Imprimer
ESD safety shoes, jo_SPEEDY, Low, S3, Jori
Fournisseur: ELTEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
12181/36 12181/37 12181/38 12181/39 12181/40 12181/41 12181/42 12181/43 12181/44 12181/45 12181/46 12181/47 12181/48
ELTN12181/36EA 60.9 EUR
ELTN12181/36 ELTN12181/37 ELTN12181/38 ELTN12181/39 ELTN12181/40 ELTN12181/41 ELTN12181/42 ELTN12181/43 ELTN12181/44 ELTN12181/45 ELTN12181/46 ELTN12181/47 ELTN12181/48
ESD safety shoes, jo_SPEEDY, Low, S3, Jori
Chaussures et bottes
Excellent wearing comfort, with a padded upper edge that protects the Achilles tendon.
  • Hydrophobised microfibre/Hydrophobized textile material
  • Fabric lining for natural ventilation
  • Padded tongue
  • Full-length inlay sole ESD
  • Non-metallic, penetration-resistant midsole
  • Rubber/EVA sole jo_DYNAMIC
  • Composite toe cap
Thanks to its excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA). Metal- and leather-free.