Imprimer
ESD safety ankle boots, DANIEL Pro, Mid, S3afety
Fournisseur: ELTEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1767901/36 1767901/37 1767901/38 1767901/39 1767902/36 1767902/37 1767902/38 1767902/39 1767903/36 1767903/37 1767903/38 1767903/39
ELTN1767901/36EA 94.9 EUR
ELTN1767901/36 ELTN1767901/37 ELTN1767901/38 ELTN1767901/39 ELTN1767902/36 ELTN1767902/37 ELTN1767902/38 ELTN1767902/39 ELTN1767903/36 ELTN1767903/37 ELTN1767903/38 ELTN1767903/39
ESD safety ankle boots, DANIEL Pro, Mid, S3afety
Chaussures et bottes
This shoe is ideally tailored to the ergonomics of female feet. Excellent wearing comfort, with a padded upper edge to protect the Achilles tendon.
  • Hydrophobised nubuck leather
  • Fabric lining for natural ventilation
  • Full, padded bellows tongue
  • Semi-orthopaedic inlay sole ESD black
  • Non-metallic, penetration-resistant midsole
  • PU/PU sole ERGO-ACTIVE 2
  • TPU scuff cap and steel toe cap
Thanks to its excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA).