Imprimer
ESD safety boots, FRANCESCO XXSG, Mid, S3
Fournisseur: ELTEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
768721/36 768721/37 768721/38 768721/39 768721/40 768721/41 768721/42 768721/43 768721/44 768721/45 768721/46 768721/47 768721/48
ELTN768721/36EA 79.9 EUR
ELTN768721/36 ELTN768721/37 ELTN768721/38 ELTN768721/39 ELTN768721/40 ELTN768721/41 ELTN768721/42 ELTN768721/43 ELTN768721/44 ELTN768721/45 ELTN768721/46 ELTN768721/47 ELTN768721/48
ESD safety boots, FRANCESCO XXSG, Mid, S3
Chaussures et bottes
Excellent wearing comfort, with an ankle-wrapping, softly padded upper edge that provides stability and grip.
  • Cowhide leather
  • Fabric lining for natural ventilation
  • Full, padded bellows tongue
  • Full-length inlay sole ARTISAN ESD
  • Non-metallic, penetration-resistant midsole
  • PU/PU sole WELLMAXX Safety Grip
  • PU toe protection
  • Steel toe cap
Thanks to its excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA). Sole core made of Infinergy® by BASF.