Imprimer
ESD safety ankle boots, lace-up, REACTION XXT Pro RFID Mid, S3
Fournisseur: ELTEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
760991/36 760991/37 760991/38 760991/39 760991/40 760991/41 760991/42 760991/43 760991/44 760991/45 760991/46 760991/47 760991/48
ELTN760991/36EA 107.9 EUR
ELTN760991/36 ELTN760991/37 ELTN760991/38 ELTN760991/39 ELTN760991/40 ELTN760991/41 ELTN760991/42 ELTN760991/43 ELTN760991/44 ELTN760991/45 ELTN760991/46 ELTN760991/47 ELTN760991/48
ESD safety ankle boots, lace-up, REACTION XXT Pro RFID Mid, S3
Chaussures et bottes
Boots feature a leather belt with hook-and-loop fastener for attaching an RFID transponder to both shoes, riveted on one side., Compatible with Activ RFID Transponder P-TAG from Borema (borema.ch) as well as shoe transponder from tbm.
  • Cowhide leather
  • Fabric lining for natural ventilation
  • Full, padded bellows tongue
  • Full-length inlay sole ESD PRO black
  • Non-metallic, penetration-resistant midsole
  • PU/PU sole WELLMAXX Trainers
  • TPU scuff cap and composite toe cap
Thanks to its excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA). Sole core made of Infinergy® by BASF.
Certifications: EN ISO 20345 S3 WR/SRC, form B