Imprimer
Conical tubes, bulk, sterile, Thermo Scientific™
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
332265 332270
NUNC332265EA 198 EUR
NUNC332265 NUNC332270
Conical tubes, bulk, sterile, Thermo Scientific™
Tubes Tubes à centrifuger
The Thermo Scientific™ Conical Centrifuge Tubes are sterile and ideal for applications requiring larger sample volumes.
  • Manufactured with high quality virgin resins
  • Sterilised by gamma irradiation per ISO 11137-2
  • Non-pyrogenic, non-cytotoxic
  • DNase/Rnase, BSE/TSE and human DNA free
Certifications: Compliant with California Prop 65