Imprimer
Reusable silica gel desiccant bags, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
F420450350 F420450500 F420451000
BELAF420450350EA 72.7 EUR
BELAF420450350 BELAF420450500 BELAF420451000
Reusable silica gel desiccant bags, SP Bel-Art
Dessiccateurs Dessicants
Convenient, easy to handle packs filled with moisture absorbing silica gel beads are available in multiple sizes to fit most desiccators.
  • Durable microfiber bag securely contains the beads eliminating the risk of leakage and is unaffected by moisture
  • Indicator dot changes from blue when dry to pink when moist to easily identify when recharging is needed
  • Saturated gel can be regenerated by microwaving the bag for 6 minutes
  • Can be regenerated repeatedly without degradation
Cdt: Supplied individually sealed in a moisture-proof pouch.