Imprimer
Recycling station billboards, Slim Jim®
Fournisseur: Rubbermaid Commercial Products
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
2007907 2007908 2007910 2007912 2007909 2007911 2007905
129-2051EA 34.5 EUR
129-2051 129-2050 129-2048 129-2046 129-2049 129-2047 129-2052
Recycling station billboards, Slim Jim®
Poubelles Conteneurs de déchets
Billboards for the Slim Jim® recycling station.
  • Provide better waste stream visibility by selecting a colour and waste stream label for your facility needs
  • Available in common recycling colours (red, yellow, green, blue, dark grey, brown and black)
  • Simple and durable execution
  • Components snap in place without the use of hardware
Compatible with Slim Jim® recycling station starter kit, Slim Jim® recycling station lid inserts, and Slim Jim® recycling station waste stream label kits.