Imprimer
Polyester tape labels for M210, M210-LAB, and M211 label printers
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
152095 152096 152097 152098 152099
BRDY152095EA 28.5 EUR
BRDY152095 BRDY152096 BRDY152097 BRDY152098 BRDY152099
Polyester tape labels for M210, M210-LAB, and M211 label printers
Étiquettes
Glossy white polyester labels.
  • Permanent acrylic adhesive
  • Rounded corners
  • Suitable for freezer and autoclave
  • For use on smooth surfaces
  • Service temperature: −70 to 110 °C
  • For use with M210 and M211 portable label printers
Aggressive permanent adhesive is heat-resistant and suitable for use in harsh environments.