Imprimer
Buchner Table-Top Funnels, 610 mm (24"), SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
H146253524 H146271610 H146220000
BELAH146253524EA 7550 EUR
BELAH146253524 BELAH146271610 BELAH146220000
Buchner Table-Top Funnels, 610 mm (24"), SP Bel-Art
Entonnoirs Entonnoirs pour la filtration Entonnoirs Büchner
Filter large volumes quickly with a polyethylene plastic Table-Top Buchner Funnel using your choice of Fritware® or Perforated filter plate.
  • Removable hose barbed connector accepts 12,7 mm (½") I.D. tube and threads into ¾" NPT bung on the funnel
  • Overall height: 330 mm (13")
  • Rim to plate: 254 mm (10")
Fritware® Porous Filter Plates supported by a multiple ring grid, are made of 6,4 mm (¼") thick HDPE, with a non-porous ring at the periphery of the plate which seals filter paper when used. Fritware® Porous Filter Plates (fixed or removable) are available in two porosities (medium, 45 - 90 µm and coarse, 90 - 130 µm). Temperature range: –20...+52 °C (–4...125 °F).
Perforated Filter Plates are made of 4,8 mm (³/₁₆") thick HDPE with 4,8 mm (³/₁₆") perforations on 11 mm (⁷/₁₆") centers. Perforated Filter Plates (fixed or removable): Fixed for coarse filtration or use with a cloth or paper filter; Removable for use in precious metals recovery. Temperature range: –20...+52 °C (–4...125 °F).