Imprimer
Buchner Table-Top Funnels, 914 mm (36"), SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
H146253536 H146263536
BELAH146253536EA 12440 EUR
BELAH146253536 BELAH146263536
Buchner Table-Top Funnels, 914 mm (36"), SP Bel-Art
Entonnoirs Entonnoirs pour la filtration Entonnoirs Büchner
Filter large volumes quickly with a polyethylene plastic Table-Top Buchner Funnel using your choice of Fritware® or Perforated filter plate.
  • Removable hose barbed connector accepts 12,7 mm (½") I.D. tube and threads into ¾" NPT bung on the funnel
  • Overall height: 350 mm (13,75")
  • Rim to plate: 318 mm (12,5")
Fritware® Porous Filter Plates supported by a multiple ring grid, are made of 6,4 mm (¼") thick HDPE, with a non-porous ring at the periphery of the plate which seals filter paper when used. Fritware® Porous Filter Plates (fixed or removable) areEn savoir plus