Imprimer
Workstation with frame to add shelves and baskets, solid or perforated top
Fournisseur: TEKNOMEK
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PTSS1300X750 PTSS1600X750 PTSS1900X750
TEKMPTSS1300X750EA 0 EUR
TEKMPTSS1300X750 TEKMPTSS1600X750 TEKMPTSS1900X750
Workstation with frame to add shelves and baskets, solid or perforated top
Mobilier Tables
Our fully welded stainless steel modular workstation is manufactured from 304-grade stainless steel and is fully customisable to include sterile wire baskets and shelves depending on your needs.
  • Fully welded 304-grade stainless steel, brushed dull polish finish
  • Hook rail system to rear ready to support baskets or shelves
  • Choose from solid or perforated top
  • 150 mm floor clearance
  • Free standing
Four stainless steel adjustable feet with non-marking rubber base.
Load capacity: 250 kg
Informations de commande: Shelf and baskets sold separately; contact your Avantor representative for more information.