Imprimer
Industrial gloves, Bamboo TwinFlex® D xg
Fournisseur: UVEX
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
6009006 6009007 6009008 6009009 6009010 6009011 6009012
112-6241EA 195 EUR
112-6241 112-6242 112-6243 112-6244 112-6245 112-6246 112-6247
Industrial gloves, Bamboo TwinFlex® D xg
Gants
The patented Bamboo TwinFlex® technology combines the best of external protection and internal comfort in one 18 gauge cut protection glove (Cut Level D).
  • Particularly light, thin and flexible with high cut protection
  • Excellent sensitivity in cut protection level D
  • 45% sustainable materials; recycled polyamide and bamboo viscose
  • Dermatologically tested for skin compatibility, free from allergenic accelerators
  • Touch screen compatible
  • Adapts precisely to fit the shape of any hand within 3 to 5 minutes of wearing
Wearers experience noticeable comfort for their skin thanks to the bamboo-viscose design. The smooth fit and high moisture absorption with a rapid action provides long lasting comfort and high wearer acceptance. High performance fibres offer a high level of protection against abrasion and cut hazards. An additional protection zone between the thumb and index finger provides double the abrasion resistance (200%) compared to the inside of the glove to increase its service life and durability. Thanks to the Xtra-Grip aqua-polymer foam coating, the Bamboo TwinFlex® D xg offers excellent grip in dry and slightly damp/oily conditions. In addition, these gloves feature a total of 45% sustainable materials: Recycled polyamide and bamboo-viscose, which is based on renewable raw materials.
Bamboo TwinFlex® D xg gloves are ideally suited for use in glass and metalworking industries, the automotive industry, as well as precision work, quality control and food processing.
Certifications: EN 388:2016 + A1:2018, EN 407:2020, ISO 21420:2020.