Imprimer
VWR®, PETG Erlenmeyer Shaker Flask, Baffled Bottom
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
214-0466EA 125 EUR
214-0466 214-0467 214-0468 214-0469 214-0470 214-0473 214-0474 214-0475
VWR®, PETG Erlenmeyer Shaker Flask, Baffled Bottom
Flasks Erlenmeyer Flasks
VWR® crystal-clear PETG Erlenmeyer shaker flasks are robust and break resistant, increasing personal safety. They are ideal for shaker culture applications and storage. The baffled bottom improves mixing and increase gas exchange.
  • Temperature range: −60 to +60 °C
  • Unlike glass, flasks will not break when dropped
  • Moulded-in graduations for accuracy
  • Vent cap option for continuous gas exchange while ensuring sterility and preventing leakage
  • Non cytotoxic, DNase-/RNase-free and non pyrogenic
  • Unique baffled design with a molded-in“1/3 Fill” line for customer convenience
  • Sterilised by irradiation SAL 10⁻⁶ (ISO 11137)
  • Shelf life: 3 years after the month of production
Manufactured in a Class 100,000 room environment under ISO 13485 and ISO 9001 quality management systems.
Informations de commande: Available with plug seal cap or vent cap made of green HDPE.
Cdt: Individually packaged.