Imprimer
Single-use bioreactor systems, BIOne
Fournisseur: DISTEK
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
2022-1001 2022-1002 2022-1014 2022-1015 2022-1018 2022-1021 2022-1024 2022-1025 2022-1028 2022-1029 2022-1005 2022-1006 2022-1016 2022-1017 2022-1019 2022-1022 2022-1023 2022-1026 2022-1027 2022-1030 2022-1031 2022-1034 2022-1035 2022-1036 2022-1038 2022-1040 2022-1056 2022-1057
DISI2022-1001EA 0 EUR
DISI2022-1001 DISI2022-1002 DISI2022-1014 DISI2022-1015 DISI2022-1018 DISI2022-1021 DISI2022-1024 DISI2022-1025 DISI2022-1028 DISI2022-1029 DISI2022-1005 DISI2022-1006 DISI2022-1016 DISI2022-1017 DISI2022-1019 DISI2022-1022 DISI2022-1023 DISI2022-1026 DISI2022-1027 DISI2022-1030 DISI2022-1031 DISI2022-1034 DISI2022-1035 DISI2022-1036 DISI2022-1038 DISI2022-1040 DISI2022-1056 DISI2022-1057
Single-use bioreactor systems, BIOne
Bioréacteurs
Distek, Inc. has developed a bench top scale single-use bioreactor (SUB) for mammalian cell growth, cell therapy, insect cells and other unique applications performed in a stirred tank bioreactor. The gamma irradiated BIOne SUB is engineered with a disposable head plate welded to a triple-layered liner that can be easily inserted into a non sterile bioreactor glass vessel, converting it to a disposable SUB within a matter of seconds.
  • Customisable above and below the head plate
  • 2 and 5 L working volume
  • Optional single-use pH probe
  • Increased throughput
  • Weldable tubing
  • Non invasive DO probe port
  • USP class VI materials
  • Animal derivative-free and low antioxidant liner material
  • Compatible with most manufacturers’ glass vessels and controllers
Upon media addition, the liner expands and moulds to the glass vessel, preserving the geometry of the existing bioreactor. The BIOne SUB’s liner material emulates the large-scale SUBs used in pilot and commercial biopharma production facilities around the world. Performing R&D studies using the most similar material on the market streamlines your tech transfer and can de-risk your process before transferring to pilot scale.
Informations de commande: Requires one BIOne starter kit (P/N 2800-0510) for each controller being converted for use with the BIOne SUB. Requires one BIOne motor adapter assembly for each controller being converted for use with the BIOne SUB. Motor adapters are controller motor-specific. For more details please contact your local Avantor sales office.