Imprimer
AmpliPur™ Expert Tips
Fournisseur: GILSON
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
F174201 F174301 F174401 F174501
732-3852EA 103 EUR
732-3852 732-3853 732-3854 732-3855
AmpliPur™ Expert Tips
Pointes de pipettes
AmpliPur™ Expert Tips are ISO 8655 compliant and provide a significant value for the money spent. They are equipped with a filter to help protect precious samples and your pipettes from cross-contamination.
  • Designed to fit various pipette brands for maximum convenience on the bench
  • Ideal for molecular biology
AmpliPur™ Expert Tips are certified DNase- and RNase-free. It makes them the preferred tool for molecular biology applications, supporting you in ensuring the limit of detection of your experiment, resulting in more sensitive nucleic acidEn savoir plus