Imprimer
9000 Safety spectacle, Stealth™
Fournisseur: JSP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ASA770-15N-800 ASA770-15P-900 ASA770-16P-000 ASA770-16R-100 ASA770-15W-900 ASA770-16X-600
JSPAASA770-15N-800EA 20.1 EUR
JSPAASA770-15N-800 JSPAASA770-15P-900 JSPAASA770-16P-000 JSPAASA770-16R-100 JSPAASA770-15W-900 JSPAASA770-16X-600
9000 Safety spectacle, Stealth™
Lunettes
The Stealth™ 9000 is a lightweight safety spectacle, weighing just 25 g, with a wraparound 9 base curve polycarbonate lens.
  • Lightweight safety spectacle weighing just 25 g
  • Adjustable nose bridge
  • UVA, UVB, and UVC protection
  • Class 1 optics with PremierShield™ (anti-scratch and anti-mist)
  • PVC nose pad
  • Polycarbonate frame
The Stealth™ 9000 safety spectacle features slim stylish temples that have been co-injection moulded with soft rubber to increase grip and provide excellent comfort levels. All of the Stealth™ 9000 safety spectacles come with class 1 optical lenses and block 99% of ultraviolet radiation. The soft nose bridge is adjustable and provides high grip fitting that prevents slippage.
Superior hard-coated class 1 optics are available in a range of shades to suit many applications.
Certifications: Conforms to EN 166 1.F.