Imprimer
VISTAshield® safety helmet, EVO®
Fournisseur: JSP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
AMC170-004-F00 AMD170-004-F00 AMD170-005-F00 AMD170-006-F00 AMD170-007-F00
JSPAAMC170-004-F00EA 85.7 EUR
JSPAAMC170-004-F00 JSPAAMD170-004-F00 JSPAAMD170-005-F00 JSPAAMD170-006-F00 JSPAAMD170-007-F00
VISTAshield® safety helmet, EVO®
Casques Casques de sécurité
EVO® VISTAshield® is a next generation, feature-rich safety helmet based on the proven Evolution® head protection technology. Incorporating a fully retractable optical class 1 integrated face shield offering A-rated impact protection. The eyewear is easy to deploy, adjust and maintain, ensuring protection is on hand whenever it is required.
  • Class 1 optical A-rated integrated face shield
  • Easy to deploy eye protection
  • Safety cell prevents ricocheting debris injuring the scalp
  • Wear prescription eyewear without compromise
  • ABS shell
  • Wheel ratchet adjustment
  • K&N rated
The EVO® VISTAshield® helmet combines a super strong ABS shell designed for superior all day protection with the comfort benefits of the Evolution® 3D-Adjustment™ harness system. Compact and lightweight at only 515 g. A fully adjustable wheel ratchet and unique 3-D adjustment secures the helmet. A 6-point terylene harness and an Egyptian cotton Chamlon™ sweatband provide unrivalled comfort.
The safety cell concept prevents ricocheting debris travelling up the eyewear and impacting the scalp. In addition the over spec cannot make contact with the scalp, preventing injury should it be forced back on to the safety cell. EVO® VISTAshield® is effortless to retract and deploy. It pivots smoothly, allowing the wearer to adjust the face shield position to maximise coverage and protection. The optical class 1 visor provides a panoramic view with minimal distortion. Conforms to EN 166.1.AT, offering impact protection at speeds of up to 425 mph (685 kph), tested using a 6 mm steel ball bearing.
Certifications: EN 397; EN 50365 Electrical Insulation.