Imprimer
Helmet mounted cough guard
Fournisseur: JSP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
JSPAAHV960-001-100EA 21.9 EUR
JSPAAHV960-001-100
Helmet mounted cough guard
Écrans de protection
Easily fits JSP EVO® safety helmets, micro peak, and standard peak variants.
  • Grade B impact protection and class 1 optical quality
  • Prevents wearer from touching eyes and face
  • Quick and simple cleaning with soap and water
  • Reliable liquid splash protection
The only JSP verified cough guard helmet fitment. Liquid splash protection helps to block aerosols and droplets. Grade B impact protection, and class 1 optical quality conforming to EN 166:2001.
Easily fits JSP EVO® safety helmets, micro peak and standard peak variants. Compatible with Sonis® ear defenders and all 2- and 4-point chinstraps.
Certifications: Conforms to EN 166:2001.