Imprimer
Elasticated chinstrap for JSP helmets
Fournisseur: JSP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
AHV001-100-000
JSPAAHV001-100-000EA 43.4 EUR
JSPAAHV001-100-000
Elasticated chinstrap for JSP helmets
Casques Casques de sécurité
Elasticated chinstrap releases automatically with forces between 150 and 250 Newtons.
  • Wide elastic chinstrap
  • Releases automatically with forces between 150 and 250 Newtons
  • Nylon and elastic webbing
  • HDPE clips
Fits all Evolution® safety helmets.
Certifications: EN 397, EN 812.