Imprimer
Model X range disposable masks, Respair®
Fournisseur: JSP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
FFP2-20 FFP2V-10 FFP3-20 FFP3V-10
JSPAFFP2-20EA 79.2 EUR
JSPAFFP2-20 JSPAFFP2V-10 JSPAFFP3-20 JSPAFFP3V-10
Model X range disposable masks, Respair®
Protection respiratoire Masques à usage unique Demi-masques fitrants contre les particules
A range of vertical fold-flat disposable masks with unique bi-directional filter material, allowing freedom of movement and easy communication.
  • Lightweight and individually wrapped
  • Ergonomically shaped for comfort and ease of use
  • Adjustable head straps provide an individual fit
  • Bi-directional filter protects the wearer and the wearer's environment
  • Valved and unvalved versions available
An easy fit minimises leakage to ensure a high level of protection and comfort, plus complete compatibility with all other forms of protective equipment for an especially clear field of vision. A range of protection factors are available in both valved and non-valved versions. FFP2 rated masks filter 94% of airborne particulates and FFP3 rated masks filter 99% of airborne particulates.
Certifications: EN149.