Imprimer
Positive pressure respirators, PowerCap® Active™
Fournisseur: JSP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
CAE602-941-100 CAU601-001-100 CAU610-000-010 CAU340-001-100 CAU330-001-100 CAU080-000-000 CAU350-000-000 CAU071-001-100 CAU150-000-000 CAU030-000-000 CAE601-941-100
JSPACAE602-941-100EA 1580 EUR
JSPACAE602-941-100 JSPACAU601-001-100 JSPACAU610-000-010 JSPACAU340-001-100 JSPACAU330-001-100 JSPACAU080-000-000 JSPACAU350-000-000 JSPACAU071-001-100 JSPACAU150-000-000 JSPACAU030-000-000 JSPACAE601-941-100
Positive pressure respirators, PowerCap® Active™
Protection respiratoire Systèmes de respiration
The PowerCap® Active™ is a lightweight positive pressure respirator with eye and face protection, as well as EN812 bumpcap protection. Designed to protect the user from dust and most airborne particulate pollutants to EN12941 TH1P.
  • Lightweight, firm fitting, ergonomically designed positive pressure respirator
  • No face fit required
  • Cap uses JSP Hardcap A1+™ bump cap technology
  • Each battery lasts 8 hours
  • Polycarbonate face shield
  • Foam face seal
PowerCap® Active™ is designed to protect the user from dust and most airborne particulate pollutants to EN 12941+A2 TH1P, while the Powercap® Active™ IP incorporates an impact faceshield with high protection against flying particles.
PowerCap® Active™ and Powercap® Active™ IP excel in very dusty environments within light industry (machining and milling), agriculture (livestock, poultry farming, grain silos), landscaping, and food processing. They offer a unique alternative to disposable masks, giving improved comfort, safety, and efficiency to the wearer. The mask gives a cooling, comfortable supply of filtered air at 160 litres per minute, eliminating breathing resistance and visor misting. The cap is easily removable and machine washable for hygiene purposes thanks to the patented connection mechanism of the cap and visor.
Eye and face impact protection to EN 166 1B, ideal for working in environments where there is a danger of flying particles. Peel-off covers are available separately. Each battery lasts 8 hours, and can also be charged while still attached to the cap. The whole unit can be conveniently sat in the charging dock, ideal for overnight charging.
Certifications: CE 682736. Respiratory performance meets the requirements of: EN 12941 TH1. Hardcap meets the requirements of EN 812. Faceshield meets the requirements of EN166 1.B.3.