Imprimer
Dust filters, PressToCheck™
Fournisseur: JSP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BMN990-001-700 BMN750-000-600 BMN740-000-600
JSPABMN990-001-700EA 28.9 EUR
JSPABMN990-001-700 JSPABMN750-000-600 JSPABMN740-000-600
Dust filters, PressToCheck™
Filtres Filtres de ventilation
PressToCheck™ twin filters for the Force®8 half mask and Force®10 full face mask facilitate instant and accurate checking of the face fit before entering the hazard area, without affecting the seal of the mask to the face.
  • Longer life/greater filter efficiency
  • Better protection in wet environments
  • Less chance of damage from foreign objects
  • Resistance to clogging from very fine dust environments
  • Improved hygiene
PressToCheck™ twin filters feature a bayonet fitting that makes the filters easy to attach. Lightweight, easy to wear, and available as dust or dust/gas combination filters. Filters resist clogging in very fine dust environments (passes Dolomite test).
Low profile cover protects the enclosed pleated filter from moisture and foreign objects that could damage the pleats. The unique design of the filter allows air to flow across the full area of the filter material and the large open area allows for lower breathing resistance and improved comfort. The filter features a built-in inhalation valve making the respirator easier to maintain.
Certifications: EN143; EN14387.