Imprimer
Horizontal fold flat disposable masks, Springfit™
Fournisseur: JSP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BGA122-202-000 BGA172-202-000 BGA132-206-000 BGA182-206-000 BGA802-206-000
JSPABGA122-202-000EA 40.6 EUR
JSPABGA122-202-000 JSPABGA172-202-000 JSPABGA132-206-000 JSPABGA182-206-000 JSPABGA802-206-000
Horizontal fold flat disposable masks, Springfit™
Protection respiratoire Masques à usage unique Demi-masques fitrants contre les particules
The SpringFit™ fold flat disposable mask achieves an unrivalled secure fit with the use of a unique endoskeleton structure. SpringFit™ introduces an abundance of features that maximise fit, comfort, compatibility, and performance.
  • Fold flat disposable mask
  • Compact and easily pocketed
  • Unique endoskeleton structure
  • Polypropylene inner skeleton
  • Available in FFP3 and FFP2, valved and unvalved
The SpringFit™ fold flat disposable mask is available in FFP3 and FFP2, valved and unvalved, with a nuisance odour carbon layer available on the FFP3 model. The Unique Typhoon™ valve has been designed to reduce breathing resistance, heat, and moisture build-up. The valve is unaffected by any moisture, and therefore can be used with the same performance under very low temperatures.
The fold flat design makes SpringFit™ more compact, and easily pocketed. SpringFit™ is also individually packed to ensure a high level of hygiene.
Certifications: EN149:2001+A1:2009.