Imprimer
PU banded ear plugs, Soundstopper™
Fournisseur: JSP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Soundstopper™
JSPAAEE090-070-21XEA 6.9 EUR
JSPAAEE090-070-21X
PU banded ear plugs, Soundstopper™
Bouchons d'oreille Arceaux anti-bruits
Lightweight and comfortable PU foam earplugs are designed to be worn banded under the chin, ideal for situations where they need to be accessible for long periods of time.
  • Lightweight and comfortable to use
  • Mould easily to achieve comfortable fit
  • SNR: 20
  • Band is adjustable for added comfort
These PU foam ear plugs can be conveniently stored around the neck when not in use. Lightweight and comfortable to use over prolonged periods. Easy to fit non-invasive design (earplugs do not penetrate the ear canal).
Certifications: Conforms to EN 352-2.