Imprimer
Cleanroom wipes, pre-saturated, PROSAT® Pi
Fournisseur: CONTEC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PS-314-BPR PS-314IR-BPR
115-2944EA 1130 EUR
115-2944 115-2945
Cleanroom wipes, pre-saturated, PROSAT® Pi
Lingettes Lingettes pour salles propres Lingettes pour salles propres, présaturées
Hydroentangled split microfibre wipes pre-saturated with 70% IPA and 30% DI water.
  • Greater surface coverage means less resource consumption including time, labour and materials
  • Pre-saturated with 70% IPA / 30% DI water; controls saturation levels which greatly reduce solvent usage, VOCs and waste
  • Non woven split microfibre increases cleaning effectiveness and contaminant pick-up
PROSAT® Pi wipes are designed to help life science manufacturers control contamination and ultimately maintain cGMP compliance while consuming less resources. The wipes are made from polyester and nylon split microfibre, enhancing their cleaning effectiveness and contaminant pick-up ability. The 80 gsm microfibre fabric is highly sorbent and offers metered release of solvent to surfaces, covering greater areas compared to other pre-saturated wipes.
PROSAT® Pi wipes are pre-saturated with 70% USP grade IPA and 30% DI water, allowing for controlled application to surfaces and less VOCs, compared to spraying. The 'peel and reseal' pouch can be resealed throughout use to maintain the wipes’ saturation level and IPA concentration. PROSAT® Pi wipes are ideal for pharmaceutical, biotechnology, and medical device manufacturers.