Imprimer
Cleanroom wipes, pre-saturated, meltblown polypropylene, PROSAT®
Fournisseur: CONTEC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PS-850-BPR PS-850E-BPR PS-911-BPR PSPP0043-BPR
115-2747EA 1170 EUR
115-2747 115-2760 115-2771 115-2816
Cleanroom wipes, pre-saturated, meltblown polypropylene, PROSAT®
Lingettes Lingettes pour salles propres Lingettes pour salles propres, présaturées
Meltblown polypropylene wipes presaturated with 70% IPA and 30% DI water.
  • Suitable for ISO Class 5-8 environments
  • Meltblown polypropylene fabric is exceptionally clean and free from additives
  • Low in particles and fibres, ideal for a variety of critical applications
  • Microsized filaments provide exceptional particle removal
  • Reduces VOC emissions and solvent use
  • Resealable pouch preserves cleanliness and solvent saturation levels
PROSAT® meltblown polypropylene wipes offer exceptional cleanliness suitable for use in a wide variety of criticalEn savoir plus