Imprimer
Removable Autoclave-Resistant Thermal Transfer Labels
Fournisseur: GA
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
AUTR-11C1-2WH AUTR-11C3-4WH AUTR-127C1-1WH AUTR-127C3-2WH AUTR-28C1-1WH AUTR-28C3-2WH AUTR-56C1-1WH AUTR-56C3-1WH AUTR61NPNO14WH AUTR61NPNO38WH AUTR-9C1-1WH AUTR-9C3-3WH
GAINAUTR-11C1-2WHEA 91.5 EUR
GAINAUTR-11C1-2WH GAINAUTR-11C3-4WH GAINAUTR-127C1-1WH GAINAUTR-127C3-2WH GAINAUTR-28C1-1WH GAINAUTR-28C3-2WH GAINAUTR-56C1-1WH GAINAUTR-56C3-1WH GAINAUTR61NPNO14WH GAINAUTR61NPNO38WH GAINAUTR-9C1-1WH GAINAUTR-9C3-3WH
Removable Autoclave-Resistant Thermal Transfer Labels
Étiquettes
These labels are intended for identifying metal, plastic, and glass surfaces, including surgical instruments, glassware, trays, blister packs, and other items that will undergo sterilisation.
  • Temperature range: −60 to 150 °C (−76 to 302 °F)
  • Glove-friendly adhesive won’t stick to gloves and ensures the labels are readily removable without leaving behind any adhesive residue
  • Can resist elevated temperatures; ideal for autoclave sterilisation cycles
  • Ideal for multiple autoclave sterilisation cycles, gamma irradiation (up to 50 KGy), and ethylene oxide sterilisation
  • Printable with all major thermal-transfer printer brands
Removable autoclave-resistant labels are intended for use in steam autoclaves, ovens, gamma irradiators, and ethylene oxide sterilisation. Durable autoclave thermal-transfer labels will not leave any adhesive residue after removal. They are weather- and water-proof, alcohol-resistant, and have been tested to endure up to 20 autoclave cycles (at 121 °C/250 °F, 16,5 psi for 20 minutes). Compatible with all major brands of thermal-transfer printers.