Imprimer
VWR®, Eco Side Access Rack for Cryoboxes, Height 50 mm
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
479-2276EA 124 EUR
479-2276 479-2277 479-2278 479-2272 479-2273 479-2274 479-2275
VWR®, Eco Side Access Rack for Cryoboxes, Height 50 mm
Portoirs Portoirs pour cryoboîtes
Freezer racks as storage and inventory systems for upright ULT freezers makes it easier organise your samples.
  • Different height and depth dimensions are available and can be used in all freezer brands
  • Racks for standard 2" cryoboxes (133×133×51 mm)
  • Made from corrosion resistant stainless steel with smoothed edges
  • Organise and find your samples quickly
  • Lower weight compared with side access racks
These racks are made from corrosion resistant stainless steel with smoothed edges. Eco Side access racks are a cost-effective freezer rack that will fit into any freezer.
Measure the inside height and depth in the compartment of your freezer before choosing the right racks. In 'Related information' tab you will find additional information to support you in the selection.
Informations de livraison: Delivered without cryoboxes.