Imprimer
Cryogenic labels for the identification of IVF straws
Fournisseur: GA
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
CST28NP.25C1WH CST28NPC3-1WH CST28NP.05C1WH CST28NP0.5C3WH CST29NPC1-1BL CST29NPC1-1GA CST29NPC1-1PI CST29NPC1-1RE CST29NPC1-1WH CST29NPC1-1YE CST29NPC3-1BL CST29NPC3-1GA CST29NPC3-1PI CST29NPC3-1RE CST29NPC3-1WH CST29NPC3-1YE CST29NP0.5C1PI CST29NP0.5C1RE CST29NP0.5C1WH CST29NP0.5C3PI CST29NP0.5C3RE CST29NP0.5C3WH CST79NPNC1-1WH CST79NPC3-1WH CST79NP0.5C1WH CST79NP0.5C3WH
GAINCST28NP.25C1WHEA 115 EUR
GAINCST28NP.25C1WH GAINCST28NPC3-1WH GAINCST28NP.05C1WH GAINCST28NP0.5C3WH GAINCST29NPC1-1BL GAINCST29NPC1-1GA GAINCST29NPC1-1PI GAINCST29NPC1-1RE GAINCST29NPC1-1WH GAINCST29NPC1-1YE GAINCST29NPC3-1BL GAINCST29NPC3-1GA GAINCST29NPC3-1PI GAINCST29NPC3-1RE GAINCST29NPC3-1WH GAINCST29NPC3-1YE GAINCST29NP0.5C1PI GAINCST29NP0.5C1RE GAINCST29NP0.5C1WH GAINCST29NP0.5C3PI GAINCST29NP0.5C3RE GAINCST29NP0.5C3WH GAINCST79NPNC1-1WH GAINCST79NPC3-1WH GAINCST79NP0.5C1WH GAINCST79NP0.5C3WH
Cryogenic labels for the identification of IVF straws
Étiquettes
Printable with thermal-transfer barcode printers for secure sample identification and traceability.
  • Self-laminating, wrap-around labels provide enhanced resistance to abrasion, chemicals, scratching, and smudging
  • For short and long-term storage in vapor or liquid phase liquid nitrogen (–196 °C/–321 °F)
  • Specifically designed for the identification of cryo-preservation straws (0,25 ml and 0,5 ml) used in IVF laboratories and clinics
  • Available in standard white, as well as an assortment of colors for quick and easy identification via color-coding
These Labels are used in assisted reproductive technology (ART) clinics and laboratories, as well as animal breeding and biotechnology companies. These durable cryo IVF straw labels can also withstand the vitrification process and are compatible with all standard cryo straws as well as high-security straws.