Imprimer
Tamper-evident and destructible cryogenic labels
Fournisseur: GA
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
TELA-2C1-1WH TELA-2C3-1WH TELA-3NPC1-1WH TELA-3NPC3-1WH TELA-5C1-1WH TELA-5C3-1WH
GAINTELA-2C1-1WHEA 133 EUR
GAINTELA-2C1-1WH GAINTELA-2C3-1WH GAINTELA-3NPC1-1WH GAINTELA-3NPC3-1WH GAINTELA-5C1-1WH GAINTELA-5C3-1WH
Tamper-evident and destructible cryogenic labels
Étiquettes
Cryo C-KurTAG™ thermoplastic waterproof cryogenic labels will tear upon any attempt to remove them.
  • Strong permanent adhesive ensures labels will not wrinkle or peel
  • Tamper-evident seal ensures any attempt to open the container will be noticed
  • Resist extreme cryogenic temperatures (–196 °C/–321 °F, in liquid or vapor phase liquid nitrogen)
Ideal for cryogenic vials and containers that require evidence of tampering when stored in liquid nitrogen or transported on dry ice. They can be used with cryogenic tubes and vials up to 5 ml in volume, freezer boxes, cryogenic boxes, as well as cell culture plates and other containers.
Sophisticated and practical design adds rigidity and strength to reduce accidental seal ruptures, yet destructs easily upon any attempt of removal. They can be printed on with thermal transfer barcode printers to include serial numbers, barcodes, and alphanumeric text.
Informations de commande: Available in different sizes and configurations.