Imprimer
Cryogenic Tamper-Evident Tapes
Fournisseur: GA
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
TETA-22C1-50WH TETA-22C1-50YE TETA-22C1-50OR TETA-22C1-50SI TETA-22C1-50GA TETA-22C1-50BL TETA-22C1-50RE TETA22C3-100WH TETA22C3-100YE TETA22C3-100OR TETA22C3-100SI TETA22C3-100GA TETA22C3-100BL TETA22C3-100RE
GAINTETA-22C1-50WHEA 36.2 EUR
GAINTETA-22C1-50WH GAINTETA-22C1-50YE GAINTETA-22C1-50OR GAINTETA-22C1-50SI GAINTETA-22C1-50GA GAINTETA-22C1-50BL GAINTETA-22C1-50RE GAINTETA22C3-100WH GAINTETA22C3-100YE GAINTETA22C3-100OR GAINTETA22C3-100SI GAINTETA22C3-100GA GAINTETA22C3-100BL GAINTETA22C3-100RE
Cryogenic Tamper-Evident Tapes
Rubans adhésifs
This tamper-evident tape is designed for use with cryogenic containers such as cryo vials, cryo/freezer boxes, cryo cassettes, and tubes.
  • Temperature range: −196 to +110 °C (−320,8 to 230 °F)
  • Able to withstand immersion in liquid nitrogen and deep freeze storage conditions
  • Continuous tape format allows use of as much or as little as necessary
  • Available in assorted colours for easy identification and colour coding
The continuous tape will tear-up upon any attempt to remove it from most surfaces, indicating whether the contents have been compromised. Durable and waterproof, the tamper-evident tape is available in a range of colours for easy identification. Writable using permanent cryogenic markers, the cryo tape is suitable for long-term storage in liquid nitrogen (−196 °C), laboratory freezers (−80 °C), and for transportation on dry ice.