Imprimer
Permanent cryogenic labels for laser printers, US letter size
Fournisseur: GA
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
CL-15T1-OR CL-15T1-PI
GAINCL-15T1-OREA 70.7 EUR
GAINCL-15T1-OR GAINCL-15T1-PI
Permanent cryogenic labels for laser printers, US letter size
Étiquettes
Cryo-LazrTAG™ laser printable cryogenic labels are designed for the permanent identification of cryo vials and tubes, microtubes, blood collection tubes, cryo boxes, freezer boxes, cell culture plates, microplates, and other cryogenic and freezer containers.
  • Strong permanent adhesive ensures labels will not wrinkle or peel
  • Printable with all common desktop laser printer brands (US Letter Size: 21,5×27,9 cm)
  • Withstand a wide temperature range (–196 to 120 °C)
  • Available in a wide assortment of colors, sizes, and configurations
These labels are designed to withstand long-term storage in vapor and liquid phase liquid nitrogen (–196 °C/–321 °F), lab freezers (–80 °C/–112 °F), and transportation on dry ice. A wide selection of white and color labels is available. Waterproof, weatherproof, and wear-and-tear proof, laser printable labels for deep-freeze and cryogenic use are also stable at elevated temperatures up to 120 °C/248 °F, and will withstand repeated freeze-thaw cycles, and gamma irradiation (tested up to 50 KGy).
CL-12 Assorted colours: white, silver, yellow, lemon, green apple, pink, lavender, blue, orange, red, mint, green seafoam
CL-23 Assorted colours: white, silver, beige, yellow, lemon, green apple, pink, lavender, blue, orange, red, mint
CL-13 Assorted colours: white, silver, yellow, lemon, green apple, pink, lavender, blue, orange, red, mint, beige
CL-6 Assorted colours: white, silver, yellow, lemon, green apple, pink, lavender, blue, orange, red, mint, beige
CL-4 Assorted colours: white, silver, yellow, lemon, green apple, pink, lavender, blue, orange, red, mint, green seafoam
CL-8 Assorted colours: white, silver, yellow, lemon, green apple, pink, lavender, blue, orange, red, moka, beige
CL-9 Assorted colours: white, silver, yellow, lemon, green apple, pink, lavender, blue, orange, red, mint, beige