Imprimer
ESD safety shoes, lace-up, Larkin XXSports Low
Fournisseur: ELTEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
728111/35 728111/36 728111/37 728111/38 728111/39 728111/40 728111/41 728111/42 728111/43 728111/44 728111/45 728111/46 728111/47 728111/48 728101/35 728101/36 728101/37 728101/38 728101/39 728101/40 728101/41 728101/42 728101/43 728101/44 728101/45 728101/46 728101/47 728101/48
ELTN728111/35EA 90.9 EUR
ELTN728111/35 ELTN728111/36 ELTN728111/37 ELTN728111/38 ELTN728111/39 ELTN728111/40 ELTN728111/41 ELTN728111/42 ELTN728111/43 ELTN728111/44 ELTN728111/45 ELTN728111/46 ELTN728111/47 ELTN728111/48 ELTN728101/35 ELTN728101/36 ELTN728101/37 ELTN728101/38 ELTN728101/39 ELTN728101/40 ELTN728101/41 ELTN728101/42 ELTN728101/43 ELTN728101/44 ELTN728101/45 ELTN728101/46 ELTN728101/47 ELTN728101/48
ESD safety shoes, lace-up, Larkin XXSports Low
Chaussures et bottes
Thanks to its excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA).
  • Padded tongue
  • TPU/PU sole WELLMAXX SPORTS
  • Steel toe cap
  • Leather-free
Mesh material construction with fabric lining for natural ventilation. Full-length inlay sole, SPORTIVE ESD. Non-metallic, penetration-resistant midsole. Sole core made of Infinergy® by BASF.
Certifications: EN ISO 20345 S1P SRA, form A.