Imprimer
ESD safety shoes, slip-on, jo_Clean Slipper Low, Jori
Fournisseur: ELTEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
12671/36 12671/37 12671/38 12671/39 12671/40 12671/41 12671/42 12671/43 12671/44 12671/45 12671/46 12671/47 12671/48 12661/36 12661/37 12661/38 12661/39 12661/40 12661/41 12661/42 12661/43 12661/44 12661/45 12661/46 12661/47 12661/48
ELTN12671/36EA 55.9 EUR
ELTN12671/36 ELTN12671/37 ELTN12671/38 ELTN12671/39 ELTN12671/40 ELTN12671/41 ELTN12671/42 ELTN12671/43 ELTN12671/44 ELTN12671/45 ELTN12671/46 ELTN12671/47 ELTN12671/48 ELTN12661/36 ELTN12661/37 ELTN12661/38 ELTN12661/39 ELTN12661/40 ELTN12661/41 ELTN12661/42 ELTN12661/43 ELTN12661/44 ELTN12661/45 ELTN12661/46 ELTN12661/47 ELTN12661/48
ESD safety shoes, slip-on, jo_Clean Slipper Low, Jori
Chaussures et bottes
ESD safety shoe made from smooth leather and fuel resistant PU sole.Thanks to its excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA).
  • Full bellows tongue
  • MONO-PU sole jo_MONO
  • Composite toe cap
  • Metal- and leather-free
  • Available in black or white finish
Hydrophobised microfibre construction with fabric lining for natural ventilation. Full-length inlay sole. Non-metallic, penetration-resistant midsole.
Certifications: EN ISO 20345 S3 SRC, form A.