Imprimer
ESD safety ankle boots, lace-up, HANNAH XXTL Mid
Fournisseur: ELTEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
746531/35 746531/36 746531/37 746531/38 746531/39 746531/40 746531/41 746531/42
ELTN746531/35EA 93.9 EUR
ELTN746531/35 ELTN746531/36 ELTN746531/37 ELTN746531/38 ELTN746531/39 ELTN746531/40 ELTN746531/41 ELTN746531/42
ESD safety ankle boots, lace-up, HANNAH XXTL Mid
Chaussures et bottes
These ESD safety ankle boots are ideally tailored to the ergonomics of female feet.
  • Full, padded bellows tongue
  • PU/PU sole WELLMAXX TRAINERS LADY
  • Steel toe cap
  • Non-metallic, penetration-resistant midsole
Hydrophobised microfibre/hydrophobised textile material, with fabric lining for natural ventilation. Full-length inlay sole, ESD PRO Lady Y black. Sole core made of Infinergy® by BASF.
Certifications: EN ISO 20345 S3 SRC, Form B.