Imprimer
ESD safety shoes, lace-up, S3, jo_Speedy Low, Jori
Fournisseur: ELTEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
12161/36 12161/37 12161/38 12161/39 12161/40 12161/41 12161/42 12161/43 12161/44 12161/45 12161/46 12161/47 12161/48 12151/36 12151/37 12151/38 12151/39 12151/40 12151/41 12151/42 12151/43 12151/44 12151/45 12151/46 12151/47 12151/48
ELTN12161/36EA 60.9 EUR
ELTN12161/36 ELTN12161/37 ELTN12161/38 ELTN12161/39 ELTN12161/40 ELTN12161/41 ELTN12161/42 ELTN12161/43 ELTN12161/44 ELTN12161/45 ELTN12161/46 ELTN12161/47 ELTN12161/48 ELTN12151/36 ELTN12151/37 ELTN12151/38 ELTN12151/39 ELTN12151/40 ELTN12151/41 ELTN12151/42 ELTN12151/43 ELTN12151/44 ELTN12151/45 ELTN12151/46 ELTN12151/47 ELTN12151/48
ESD safety shoes, lace-up, S3, jo_Speedy Low, Jori
Chaussures et bottes
Excellent wearing comfort, and suitable for work in ESD sensitive or electrostatically protected areas.
  • Padded tongue
  • Non-metallic, penetration-resistant midsole
  • Rubber/EVA sole jo_DYNAMIC
  • Rubber scuff cap
  • Composite toe cap
Hydrophobised textile material with fabric lining for natural ventilation. Full-length inlay ESD sole. Metal-free and leather-free.
Certifications: EN ISO 20345 S3 HRO/SRC, form A.